Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Trådnät 150 mesh woven from 0.066mm (0.0026in) Type 304
Leverantör: Thermo Fisher Scientific
42010.RF
42010.RU
42010.RFEA
3600
SEK
42010.RF
42010.RU
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Trådnät 150 mesh woven from 0.066mm (0.0026in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Wire Cloth
MDL No.: MFCD00148499