Polyethylene glycol monomethyl ether, M.W. 1900
Leverantör: Thermo Fisher Scientific
Synonyms: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH
41563.22 41563.36
41563.22EA 1425 SEK
41563.22 41563.36
Polyethylene glycol monomethyl ether, M.W. 1900
Polyethylene glycol monomethyl ether
CAS No.: 9004-74-4 Molecular Formula: CH3(OCH2CH2)nOH Flash Point: 110°(230°F) Physical Form: Solid mass Appearance: White to pale yellow Melting Point: 52° MDL No.: MFCD00084416
Formel: CH₃(OCH₂CH₂)nOH
Smältpunkt: 52 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00084416
CAS nummer: 9004-74-4