Polyethylene glycol monomethyl ether, M.W. 750
Leverantör: Thermo Fisher Scientific
41562.22 41562.36
41562.22EA 1200 SEK
41562.22 41562.36
Polyethylene glycol monomethyl ether, M.W. 750
Polyethylene glycol monomethyl ether
CAS No.: 9004-74-4 Molecular Formula: CH3(OCH2CH2)nOH Flash Point: 110°(230°F) Physical Form: Low melting solid Density: 1.10 Appearance: Colourless to pale yellow Melting Point: 28-32° MDL No.: MFCD00084416
Formel: CH₃(OCH₂CH₂)nOH
Smältpunkt: 28…32 °C
Densitet: 1,10
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00084416
CAS nummer: 9004-74-4