Chloropentaammineiridium(III) chloride (≥49,6% Ir) ≥99.9%

Leverantör: Thermo Scientific
Varning

41006.03 41006.06
41006.03EA 9475 SEK
41006.03 41006.06
Chloropentaammineiridium(III) chloride (≥49,6% Ir) ≥99.9%
Chloropentaammineiridium(III) chloride

Purity: 99.9% CAS No.: 15742-38-8 Molecular Formula: H15Cl3IrN5 Formula Weight: 383.73 Physical Form: Powder Appearance: White to pale yellow Melting Point: dec. MDL No.: MFCD00798542

Formel: H₁₅Cl₃IrN₅
MW: 383,73 g/mol
Smältpunkt: Dec.
Storage Temperature: Omgivande
MDL Number: MFCD00798542
CAS nummer: 15742-38-8
EINECS: 239-833-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR