Dihydrogen hexafluorotitanate 60% (w/w) i vattenlösning
Leverantör: Thermo Scientific
Fara
Synonyms: Hexafluorotitanic acid, Hydrogen hexafluorotitanate, Fluorotitanic acid
39736.36 39736.A1
39736.36EA 2950 SEK
39736.36 39736.A1
Dihydrogen hexafluorotitanate 60% (w/w) i vattenlösning
Dihydrogen hexafluorotitanate
CAS No.: 17439-11-1 Molecular Formula: H2TiF6 Formula Weight: 163.91 Physical Form: Liquid Density: 1.69 Odour: Pungent UN No.: UN2922 MDL No.: MFCD00049647
Formel: H₂TiF₆
MW: 163,91 g/mol
Densitet: 1,69
Storage Temperature: Omgivande
MDL Number: MFCD00049647
CAS nummer: 17439-11-1
EINECS: 241-460-4
UN: 2922
ADR: 8,II