Tin(IV) 2-ethylhexanoate diisopropoxide ≥98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
39169.03
39169.06
39169.03EA
885
SEK
39169.03
39169.06
Tin(IV) 2-ethylhexanoate diisopropoxide ≥98%
Tin(IV) 2-ethylhexanoate diisopropoxide
Purity: 98%
CAS No.: 288374-32-3
Molecular Formula: C22H44O6Sn
Formula Weight: 523.08
Physical Form: Liquid
Sensitivity: Moisture Sensitive
MDL No.: MFCD00210604
Formel:
C₂₂H₄₄O₆Sn MW: 523.08 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00210604 CAS nummer: 288374-32-3 |