Palladium 1% on activated carbon powder wetted_50%_Water, eggshell, reduced
Leverantör: Thermo Fisher Scientific
38527.14 38527.22 38527.06
38527.14EA 4975 SEK
38527.14 38527.22 38527.06
Palladium 1% on activated carbon powder wetted_50%_Water, eggshell, reduced
Palladium
CAS No.: 7440-05-3 Physical Form: 900-1100m2/g MDL No.: MFCD03457879
Formel: Pd
MW: 106.420 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3