Palladium 20% on activated carbon powder standard wetted_50%_Water, unreduced
Leverantör: Thermo Fisher Scientific
38309.06 38309.14 38309.22
38309.06EA 8000 SEK
38309.06 38309.14 38309.22
Palladium 20% on activated carbon powder standard wetted_50%_Water, unreduced
Palladium
Physical Form: 900-1100 m²/g Odour: Odourless
Beställningsinformation: Sold on a dry weight basis. Unit weight excludes water weight
Formel: Pd
MW: 106.420 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3