Aluminium oxide activated basic ≥99.9995% Brockmann Grade I 58 angstroms
Leverantör: Thermo Scientific
11503.30 11503.A7
11503.30EA 1350 SEK
11503.30 11503.A7
Aluminium oxide activated basic ≥99.9995% Brockmann Grade I 58 angstroms
Aluminium oxide activated basic
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: -60 Mesh Powder, S.A. 150m2/g Odour: Odourless Melting Point: 2045° MDL No.: MFCD00003424
Suggested for the separation of basic amino acids, amines and other basic substances. Similar to neutral alumina, it will also adsorb hydrocarbons, aromatics and similar unsaturated compounds from organic solvents
Formel: Al₂O₃
MW: 101,96 g/mol
Smältpunkt: 2045 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356