Cobalt(II) phosphate octahydrate
Leverantör: Thermo Fisher Scientific
Fara
23111.18 23111.30
23111.18EA 2425 SEK
23111.18 23111.30
Cobalt(II) phosphate octahydrate
Cobalt(II) phosphate octahydrate
CAS No.: 10294-50-5 Molecular Formula: Co3(PO4)2.8H2O Formula Weight: 510.87 (366.74anhy) Physical Form: Powder Density: 2.769 Odour: Odourless MDL No.: MFCD00016028
Formel: Co₃(PO₄)₂·8H₂O
MW: 366.74 g/mol (anhy)
Densitet: 2.769
Storage Temperature: Omgivande
MDL Number: MFCD00016028
CAS nummer: 10294-50-5
EINECS: 236-655-6