1-Cyclohexene-1-acetic acid
Leverantör: Thermo Fisher Scientific
|
Varning
|
19462.03
19462.06
19462.14
19462.03EA
940
SEK
19462.03
19462.06
19462.14
1-Cyclohexene-1-acetic acid
1-Cyclohexene-1-acetic acid
CAS No.: 18294-87-6
Molecular Formula: C8H12O2
Formula Weight: 140.18
Physical Form: Liquid/low melting solid
Density: 1.04
Refractive Index: 1.4775
Melting Point: 33°
Boiling Point: 140-142°/25mm
MDL No.: MFCD00015482
Formel:
C₈H₁₂O₂ Kokpunkt: 140…142 °C (25 mmHg) Smältpunkt: 33 °C Densitet: 1,04 Storage Temperature: Omgivande |
MDL Number:
MFCD00015482 CAS nummer: 18294-87-6 EINECS: 242-172-1 |