Lead(II) zirconate ≥99.7% (max. 1% Hf)
Leverantör: Thermo Fisher Scientific
Fara
14682.22 14682.36
14682.22EA 4950 SEK
14682.22 14682.36
Lead(II) zirconate ≥99.7% (max. 1% Hf)
Lead(II) zirconate
Purity: 99.7% CAS No.: 12060-01-4 Molecular Formula: PbZrO3 Formula Weight: 346.42 Physical Form: -325 Mesh Powder Density: 7.0 Odour: Odourless UN No.: UN3077 MDL No.: MFCD00016275
Formel: O₃PbZr
MW: 346.421 g/mol
Densitet: 7.0
Storage Temperature: Omgivande
MDL Number: MFCD00016275
CAS nummer: 12060-01-4
EINECS: 235-039-4
UN: 3077
ADR: 9 III