Samarium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
13177.14
13177.22
13177.14EA
1625
SEK
13177.14
13177.22
Samarium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Samarium(III) carbonate hydrate
Purity: 99.99%
CAS No.: 38245-37-3
Molecular Formula: Sm2(CO3)3.xH2O
Formula Weight: 480.73(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point:
Formel:
Sm₂(CO₃)₃·xH₂O MW: 480,73 g/mol (anhy) Smältpunkt: >500 °C (dec.) Storage Temperature: Omgivande |
MDL Number:
MFCD00150444 CAS nummer: 38245-37-3 EINECS: 227-581-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...