Dicobalt octacarbonyl stabilised
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
Di-Cobalt Octacarbonyl, Cobalt carbonyl (Dicobalt octacarbonyl), Octacarbonyldicobalt, Cobalt Carbonyl, Cobalt carbonyl
Stabilization:
1-5% Stabilised with hexane
CAS No.: 10210-68-1
Molecular Formula: C8Co2O8
Formula Weight: 341.95
Flash Point: -13°(-9°F)
Physical Form: Crystalline
Density: 1.73
Sensitivity: Air Sensitive
Appearance: Red-orange
Melting Point: ca 51° dec.
Solubility: Insoluble in water. Soluble in alcohol, ether and carbon disulfide
UN No.: UN3124
MDL No.: MFCD00016024
Applications: As a catalyst in oxo process, and in high-purity cobalt salts
Formel:
C₈Co₂O₈ MW: 341.95 g/mol Smältpunkt: ∼ 51 °C (dec.) Densitet: 1.73 Flampunkt: –13 °C (9 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00016024 CAS nummer: 10210-68-1 EINECS: 233-514-0 UN: 3124 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...