Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Tris(2,2'-bipyridyl)dichlororuthenium(II), Dichlorotris(2,2'-bipyridyl)ruthenium(II), Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, Tris(2,2-bipyridyl)ruthenium(II) chloride hexahydrate
12783.02 12783.04
12783.02EA 2375 SEK
12783.02 12783.04
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate ≥98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate
Purity: 98+% CAS No.: 50525-27-4 Molecular Formula: C30H24Cl2N6Ru.6H2O Formula Weight: 748.63 (640.54anhy) Physical Form: Crystalline Appearance: Red-orange Melting Point: 63-66° MDL No.: MFCD00149670
Formel: C₃₀H₃₆Cl₂N₆O₆Ru
MW: 748,62 g/mol
Smältpunkt: 63…66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149670
CAS nummer: 50525-27-4
EINECS: 238-266-7