Sodium hexanitritocobaltate(III) ACS
Leverantör: Thermo Scientific
Fara
Synonyms: Trinatriumhexanitritokobaltat
11667.22 11667.36
11667.22EA 2625 SEK
11667.22 11667.36
Sodium hexanitritocobaltate(III) ACS
Sodium hexanitritocobaltate(III)
CAS No.: 14649-73-1 Molecular Formula: Na3Co(NO2)6 Formula Weight: 403.94 Physical Form: Powder Appearance: Yellow to orange Odour: Odourless Melting Point: 220° dec. Solubility: Very soluble in water UN No.: UN1479 MDL No.: MFCD00003512
Formel: Na₃Co(NO₂)₆
MW: 403,94 g/mol
Smältpunkt: 220 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00003512
CAS nummer: 14649-73-1
EINECS: 237-077-7
UN: 1479
ADR: 5.1,II
Merck Index: 14,8603