Yttrium(III) acetate tetrahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
11186.14 11186.22
11186.14EA 2300 SEK
11186.14 11186.22
Yttrium(III) acetate tetrahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Yttrium(III) acetate tetrahydrate
Purity: 99.99% CAS No.: 85949-60-6 Molecular Formula: C6H9O6Y.4H2O Formula Weight: 338.10 (266.04anhy) Physical Form: Crystalline Aggregates Density: 1.5 Appearance: White Melting Point: dec. MDL No.: MFCD00013048
Formel: C₆H₉O₆Y·4H₂O
MW: 338.10 g/mol (266.04 anhy)
Smältpunkt: Dec.
Densitet: 1.5
Storage Temperature: Omgivande
MDL Number: MFCD00013048
CAS nummer: 85949-60-6
EINECS: 245-612-0