Tetracarbonyldi-µ-chlorodirhodium(I) (50,1 - 52,9% Rh)
Leverantör: Thermo Fisher Scientific
|
Varning
|
Synonyms:
Di-μ-chloro-tetracarbonyldirhodium(I), Chlorodicarbonylrhodium(I) dimer
10547.01
10547.03
10547.01EA
4175
SEK
10547.01
10547.03
Tetracarbonyldi-µ-chlorodirhodium(I) (50,1 - 52,9% Rh)
Tetracarbonyldi-µ-chlorodirhodium(I)
CAS No.: 14523-22-9
Molecular Formula: C4Cl2O4Rh2
Formula Weight: 388.76
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: Red
Melting Point: 121°
MDL No.: MFCD00135610
Formel:
C₄Cl₂O₄Rh₂ Smältpunkt: 121 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00135610 CAS nummer: 14523-22-9 EINECS: 238-540-6 Merck Index: 14,8187 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...