Natrium 40% dispersion in oil , 99% Metaller basis
Leverantör: Thermo Fisher Scientific
Fara
10340.30 10340.56
10340.30EA 3550 SEK
10340.30 10340.56
Natrium 40% dispersion in oil , 99% Metaller basis
Natrium
Purity: 99+% CAS No.: 7440-23-5 Sensitivity: Air and Moisture Sensitive Appearance: White to off-white UN No.: UN1391 MDL No.: MFCD00085307
Formel: Na
MW: 22.99 g/mol
Kokpunkt: 881
Smältpunkt: 97.8
Storage Temperature: Omgivande
MDL Number: MFCD00085307
CAS nummer: 7440-23-5
EINECS: 231-132-9
UN: 1391
ADR: 4.3 I
Merck Index: 13,08641