Buffer solutions, according to EP and USP, Certipur®
Leverantör: Merck
1.09406.0500
1.99011.0001
1.99015.0500
1.99017.0001
1.99068.0500
1.99091.0500
1.09406.0500EA
565
SEK
1.09406.0500
1.99011.0001
1.99015.0500
1.99017.0001
1.99068.0500
1.99091.0500
Buffer solutions, according to EP and USP, Certipur®
Buffertar
pH buffertar
Buffer solutions intended for extremely precise calibration, qualification and monitoring of pH instruments, and for testing other buffer solutions.
- Measured using 5-point calibration against NIST & PTB primary certified reference materials
- Detailed certificate of analysis for every product
- Solutions in packs of single use sachets (30×30 ml) help prevent contamination