Hexaklorplatinasyra, GR analytiskt reagens, Supelco®
Leverantör: Merck
Fara
Synonyms: Hydrogen hexachloroplatinate (IV), Dihydrogen hexachloroplatinate (IV)
M-Clarity™ quality level = MQ100
1.07341.0005 1.07341.0025
1.07341.0005EA 4540 SEK
1.07341.0005 1.07341.0025
Hexaklorplatinasyra, GR analytiskt reagens, Supelco®
Hexaklorplatinasyra
Formel: H₂Cl₆Pt
MW: 409,81 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011330
CAS nummer: 16941-12-1
UN: 3264
ADR: 8,II
Order Now
Specifikation testresultat
Solubility Soluble at 20 °C
Density 1.06 g/cm3 (20 °C)
pH 0.6 (H2O, 20 °C)
Assay ≥10,0 %