Stereo microscopes, Stemi 508
Leverantör: ZEISS
4950090006000 4950090007000 4950090008000
630-2632EA 48175 SEK
630-2632 630-2821 630-2822
Stereo microscopes, Stemi 508
Mikroskop
The Stemi 508 are designed for quality images in biomedical (IVF), botany (plants, insects) and forensics (dissection, fossiles) applications.
  • Large object field for details (36 mm)
  • Contrast and colour reproducibility
  • Robust design with camera output for outstanding three dimensional images
Beställningsinformation: Eyepieces are included.
Leveransinformation: Supplied with a VisiLED ring light.
Order Now
SPECIFIKATIONER
Modell Stemi 508 Edu Stemi 508 Doc Stemi 508 Doc
Environmentally Preferable
Okular WF 10×/23 mm WF 10×/23 mm WF 10×/23 mm
Förstoring 13× - 100× 6,5× - 50× 13× - 100×
Zoomobjektiv 8:1 8:1 8:1
Fokussystem Grov- och fininställning Grov- och fininställning Grov- och fininställning
Belysning LED LED LED