Cooling thermostats, Proline Edition X
Leverantör: LAUDA
L001603 L001604 L001605 L001606 L001611 L001612 L001613 L001614 L001619 L001620 L001621 L001622 L001618 L001623 L001624 L001625 L001610 L001615 L001616 L001617
461-1284EA 90125 SEK
461-1284 461-1285 461-1286 461-1287 LAUDL001611 LAUDL001612 LAUDL001613 LAUDL001614 LAUDL001619 LAUDL001620 LAUDL001621 LAUDL001622 LAUDL001618 LAUDL001623 LAUDL001624 LAUDL001625 LAUDL001610 LAUDL001615 LAUDL001616 LAUDL001617
Cooling thermostats, Proline Edition X
Termostater Termostatbad, slutna system Cirkulationskylare
The Proline Edition X range of cooling thermostats feature a wide temperature range, high cooling and heating power and a multi level pump. The self-explanatory menu-guided remote control Command is particularly convenient for operating the units at distances as far as 50 meters away. Thanks to the large display, the user can keep an eye on all the important parameters. There is a graphical display of measured values, large choice of interfaces (e.g. Ether- CAT, Ethernet, RS-232/485, analog module, Profibus, contact modules). Data collection together with documentation and control is easy using the Wintherm Plus software. Units are ideal for temperature control of chemical reactors, distillation plants and glass condensers, crystallisation, thermostating measurement setups in process technology, chemical probes, calibration of sensors and testing electronic components.
  • Easy to use with simple, intuitive control
  • PowerAdapt system provides maximum heating performance, up to 3,5 kW depending on the network in use
  • SmartCool system gives to 75% energy savings thanks to digital refrigeration management
  • Powerful Varioflex pump (pressure-suction pump) with eight pump levels
  • Flexible system with various interface modules, lateral and rear pump connections
  • Multifunctional: Simultaneous connection of two external applications is possible
Max. flow rate / max. pressure:
25 L/min pressure and 23 L/min suction
0,7 bar pump pressure and 0,4 bar suction performance
Certifikat: IP protection class according to IEC529; IP 21.
Leveransinformation: Supplied with Wintherm Plus software, a wide selection of accessories are available these must be ordered separately.
Observera: Cooling capacity at 20 °C.