Test sieves with beechwood frame
Leverantör: NEXOPART
206171689 206171771 206171832 206171894 206171955
HAVE206171689EA 3225 SEK
HAVE206171689 HAVE206171771 HAVE206171832 HAVE206171894 HAVE206171955
Test sieves with beechwood frame
Siktar Siktar, test
These sieves are recommended for hand sieving of coarser materials. They are used to analyse and separate great sample quantities of industrial rocks and minerals, cereals, glass, ceramic.
  • High degree of ability through strong construction
  • Not as heavy as comparable stainless steel version
Beechwood frame with stainless steel woven wire cloth
The Haver UWL 400 H was designed with special appliance and clamping system to hold test sieves with beechwood frames.
Certifikat: According to ISO 565, DIN ISO 3310, ASTM E 11, BS 410, AFNOR, NEN 2560, ISO 5223 etc.