1-(2-Pyridylazo)-2-naphthol (PAN) solution_SC 0,1% Indicator
Leverantör: Hach
Fara
Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol
2122432 2122426 2122449
HACH2122432EA 625 SEK
HACH2122432 HACH2122426 HACH2122449
1-(2-Pyridylazo)-2-naphthol (PAN) solution_SC 0,1% Indicator
1-(2-Pyridylazo)-2-naphthol
For determination of low level manganese by the PAN method.
Formel: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Kokpunkt: 412
Storage Temperature: Omgivande
MDL Number: MFCD00004071
CAS nummer: 85-85-8
UN: 3082
ADR: 9 III
Order Now
Specifikation testresultat
Method 8149
Method Name PAN
Parameter Manganese
Platform Solution
Range 0.006 - 0.700 mg/L Mn