Clamps, three pronged
Leverantör: R L ENTERPRISES
1020 1020/1
RLAA1020EA 1300 SEK
RLAA1020 RLAA1020/1
Clamps, three pronged
Klämmor Universalklämmor
Nickel plated diecast aluminium alloy. Three prong clamp with nickel plated brass pummels and blue plastic headed nickel plated steel twin screw pivoting on a stainless steel pin.
  • Jaws coated with chemical resistant plastic
  • Spring loaded jaws (only RLAA1020/1)
  • Clamp is designed to hold articles within the jaws
  • For use with articles up to 90 mm diameter