Disposable filters, Impact® SR LHVD
Leverantör: ENTEGRIS
ATFZ00001 ATFV00001 ATFG00001
ENTEATFZ00001EA 0 SEK
ENTEATFZ00001 ENTEATFV00001 ENTEATFG00001
Disposable filters, Impact® SR LHVD
Filter Filterpatroner
These disposable filters have been optimised for use with the IntelliGen® photochemical dispense system to deliver excellent filtration and dispense characteristics.
  • Compatible with aggressive solvents
  • Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
  • For use with the IntelliGen® photochemical dispense system and Impact® ST manifold
  • For small to mid-volume point of use aggressive solvent filtration applications
Membrane: PTFE; Supports, core, sleeve and shell: PFA
Maximum operating temperature: 30 °C
Maximum inlet pressure: 3,9 bar at 25 °C
Maximum forward/reverse differential pressure: 2,9 bar at 25 °C