Meker burners
Leverantör: R-L ENTERPRISES LTD
4101/21 4101/3 4101/31 4101/32 4101/4 4101/41 4101/5 4101/51 4101/52
RLAA4101/21EA 2850 SEK
RLAA4101/21 RLAA4101/3 RLAA4101/31 RLAA4101/32 RLAA4101/4 RLAA4101/41 RLAA4101/5 RLAA4101/51 RLAA4101/52
Meker burners
Värmeapparater Brännare, gas
Nickel plated brass. The top of the burner is fitted with a perforated steel head, which gives multiple small but very hot flames. It is not fitted with an air regulator. Burners have diecast base painted in a blue chemically resistant acrylic finish.