Bottles, UN, narrow neck, square, HDPE, with screw caps
Leverantör: BURKLE
0304-0500 0304-1000 0304-2500
BURK0304-0500EA 90 SEK
BURK0304-0500 BURK0304-1000 BURK0304-2500
Bottles, UN, narrow neck, square, HDPE, with screw caps
Flaskor Flaskor, plast
HDPE, translucent, with black PP screw cap.
  • Absolutely leakproof
  • With UN approval for liquids
  • Complete with tamper-evident closure with PTFE seal
The bottles are excellent for packaging purposes, sampling, sending samples, storage and shipping via the postal service.