Vatten, Cell Culture Grade pyrogen-free
Leverantör: BIOWEST
Synonyms: Hydrogen oxide
L0970-500 L0970-1000 L0970-100
L0970-500EA 100 SEK
L0970-500 L0970-1000 L0970-100
Vatten, Cell Culture Grade pyrogen-free
Vatten
Cell culture water is high-quality water suitable for use as a solvent in the preparation of cell culture media and laboratory reagents.
  • Pyrogen-free
Formel: H₂O
MW: 18,02 g/mol
Kokpunkt: 100 °C (1013 hPa)
Smältpunkt: 0 °C
Densitet: 1 g/cm³ (4 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011332
CAS nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098