6-Hydroxy-1-tetralone ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

H1335-5G H1335-25G
TCIAH1335-5GEA 805 SEK
TCIAH1335-5G TCIAH1335-25G
6-Hydroxy-1-tetralone ≥98.0% (by GC, titration analysis)
6-Hydroxy-1-tetralone
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
MDL Number: MFCD00156670
CAS nummer: 3470-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c0956e4854c8e0367f50c4d79a4053b0099d2ca&vid=3c0956e4854c8e0367f50c4d79a4053b0099d2ca&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR