Methylstyrene (mixture of isomers) ≥98.0% stabilised
Leverantör: TCI EUROPE
Fara
Stabilization: med 4-tert-Butylpyrokatekol
V0028-25ML V0028-500ML
TCIAV0028-25MLEA 230 SEK
TCIAV0028-25ML TCIAV0028-500ML
Methylstyrene (mixture of isomers) ≥98.0% stabilised
Methylstyrene (mixture of isomers)
Formel: C₉H₁₀
MW: 118,18 g/mol
Kokpunkt: 173°C
Smältpunkt: -77°C
Flampunkt: 53°C
Storage Temperature: Kylskåp
MDL Number: MFCD00168066
CAS nummer: 25013-15-4
EINECS: 246-562-2
UN: 2618
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b3cce2b4fc90dd48f7001180cd518ffc7ae8ba7&vid=7b3cce2b4fc90dd48f7001180cd518ffc7ae8ba7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)