4-Bromophenethylamine 98%
Leverantör: Thermo Scientific
|
Fara
|
Synonyms:
2-(4-Bromophenyl)ethylamine
H63706.03
H63706.06
H63706.03EA
1200
SEK
H63706.03
H63706.06
4-Bromophenethylamine 98%
4-Bromophenethylamine
Purity: 98%
CAS No.: 73918-56-6
Molecular Formula: C8H10BrN
Formula Weight: 200.08
Storage Temperature: 2°C to 8°C
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.290
Refractive Index: 1.5740
Appearance: Colourless
Boiling Point: 63-72°/0.2mm
UN No.: 2735
MDL No.: MFCD00008189
Formel:
C₈H₁₀BrN MW: 200,08 g/mol Kokpunkt: 63…72 °C (0,2 mmHg) Densitet: 1,290 Flampunkt: 113 °C (235 °F) |
MDL Number:
MFCD00008189 CAS nummer: 73918-56-6 EINECS: 277-636-2 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...