1-Hexyl-3-methylimidazolium iodide ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-hexyl-3-methylimidazolium iodide
H59487.06 H59487.18
H59487.06EA 925 SEK
H59487.06 H59487.18
1-Hexyl-3-methylimidazolium iodide ≥98%
1-Hexyl-3-methylimidazolium iodide
Formel: C₁₀H₁₉IN₂
MW: 294,18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12964976
CAS nummer: 178631-05-5