1-Hexanethiol ≥96.0%

Leverantör: TCI
Fara

Synonyms: n-Hexyl mercaptan, n-Hexanethiol, 1-Mercaptohexane

H0103-25ML H0103-500ML
TCIAH0103-25MLEA 230 SEK
TCIAH0103-25ML TCIAH0103-500ML
1-Hexanethiol ≥96.0%
1-Hexanethiol
Formel: C₆H₁₄S
Kokpunkt: 152°C
Smältpunkt: –81°C
Densitet: 0,85911 g/cm³ (0 °C)
Flampunkt: 30°C
Storage Temperature: Omgivande
MDL Number: MFCD00004909
CAS nummer: 111-31-9
EINECS: 203-857-0
UN: 1228
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9322e36027859355026e31f23a4fe6f427152a6b&vid=fcf8db7fa4ccb4009254ce6721dbf6f07eb991cc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR