3-Fluorophenyl acetate ≥98.0%

Leverantör: TCI

Synonyms: 1-Acetoxy-3-fluorobenzene

F0800-5G F0800-25G
TCIAF0800-5GEA 365 SEK
TCIAF0800-5G TCIAF0800-25G
3-Fluorophenyl acetate ≥98.0%
3-Fluorophenyl acetate
Formel: C₈H₇FO₂
Kokpunkt: 78°C
Flampunkt: 75°C
MDL Number: MFCD00142578
CAS nummer: 701-83-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3985aea0d9f86d249c87a7b92b37e4d986737837&vid=3985aea0d9f86d249c87a7b92b37e4d986737837&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR