Methyl (methylthio)acetate ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(Methylthio)acetic methyl ester
M2296-5G
M2296-25G
TCIAM2296-5GEA
525
SEK
TCIAM2296-5G
TCIAM2296-25G
Methyl (methylthio)acetate ≥99.0%
Methyl (methylthio)acetate
Formel:
C₄H₈O₂S MW: 120,17 g/mol Kokpunkt: 60°C Densitet: 1,121 g/cm³ (20 °C) Flampunkt: 58°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00039809 CAS nummer: 16630-66-3 EINECS: 240-683-4 UN: 3272 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...