2,5-Dibromo-3-nitropyridine ≥98%
Leverantör: Thermo Fisher Scientific
Varning
H61961.14 H61961.22
H61961.14EA 4850 SEK
H61961.14 H61961.22
2,5-Dibromo-3-nitropyridine ≥98%
2,5-Dibromo-3-nitropyridine
Formel: C₅H₂Br₂N₂O₂
MW: 281.89 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09266223
CAS nummer: 15862-37-0