3-Phenylpiperidine ≥98.0%

Leverantör: TCI EUROPE
Varning

P2006-5G P2006-1G
TCIAP2006-5GEA 4125 SEK
TCIAP2006-5G TCIAP2006-1G
3-Phenylpiperidine ≥98.0%
3-Phenylpiperidine
Formel: C₁₁H₁₅N
MW: 161,25 g/mol
Kokpunkt: 126°C
Smältpunkt: 15°C
Storage Temperature: Kylskåp
MDL Number: MFCD00804860
CAS nummer: 3973-62-4
EINECS: 223-602-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8529d4a6bf8174cf7ae949a782a88a3bf5d2073a&vid=8529d4a6bf8174cf7ae949a782a88a3bf5d2073a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR