D-(-)-N-Ethylglucamine ≥98.0%

Leverantör: TCI EUROPE

Synonyms: N-Ethyl-D-glucamine, (-)-1-Deoxy-1-(ethylamino)-D-glucitol, (-)-1-Ethylamino-1-deoxy-D-glucitol, (2R,3R,4R,5S)-6-(Ethylamino)hexane-1,2,3,4,5-pentaol, (-)-N-Ethyl-D-glucamine

E0923-250G E0923-25G
TCIAE0923-250GEA 1100 SEK
TCIAE0923-250G TCIAE0923-25G
D-(-)-N-Ethylglucamine ≥98.0%
D-(-)-N-Ethylglucamine
Formel: H₃CCH₂NHCH₂(CH(OH))₄CH₂OH
MW: 209,24 g/mol
MDL Number: MFCD03789564
CAS nummer: 14216-22-9
EINECS: 238-073-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf8452b324baf6ea25f0ddb88d7edb967fc280bb&vid=bf8452b324baf6ea25f0ddb88d7edb967fc280bb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 137.0 to 141.0 deg-C
Specific rotation [a]20/D -15.0 to -18.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR