Melamin monomer ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine
T0337-500G T0337-25G
TCIAT0337-500GEA 295 SEK
TCIAT0337-500G TCIAT0337-25G
Melamin monomer ≥98.0% (by HPLC, titration analysis)
Melamin
Formel: C₃H₆N₆
MW: 126,12 g/mol
Densitet: 1,57 g/cm³ (20 °C)
Flampunkt: ≥ 280 °C
MDL Number: MFCD00006055
CAS nummer: 108-78-1
EINECS: 203-615-4
Merck Index: 14,05811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=096489abad01310cdf0251353fd7b3e9dc02d5ca&vid=50ed378d79a3049fc269e73a40809d4a2cf7e4bb&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Water almost transparency