3-Chloro-4-(trifluoromethoxy)aniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
C2528-5G C2528-25G
TCIAC2528-5GEA 565 SEK
TCIAC2528-5G TCIAC2528-25G
3-Chloro-4-(trifluoromethoxy)aniline ≥98.0% (by GC, titration analysis)
3-Chloro-4-(trifluoromethoxy)aniline
Formel: C₇H₅ClF₃NO
MW: 211,57 g/mol
Kokpunkt: 241 °C (1013 hPa)
Smältpunkt: 29…30 °C
Densitet: 1,454 g/cm³ (20 °C)
MDL Number: MFCD00190126
CAS nummer: 64628-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75485df5882e97b4de22f497555be47c9ced935d&vid=75485df5882e97b4de22f497555be47c9ced935d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %