Flourescein dinatriumsalt
Leverantör: TCI
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate
F0096-25G F0096-500G F0096-100G
TCIAF0096-25GEA 215 SEK
TCIAF0096-25G TCIAF0096-500G TCIAF0096-100G
Flourescein dinatriumsalt
Flourescein dinatriumsalt
Formel: C₂₀H₁₀Na₂O₅
Smältpunkt: > 300 °C
Densitet: 1,601 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00167039
CAS nummer: 518-47-8
EINECS: 208-253-0
Merck Index: 14,04159
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e0f87df032e8949e48b76f7e79b205d17cc9a93&vid=dd2dc9e704eeef0d55f25d2692d323af2c03acba&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Amber to Brown powder to crystal
Lambda max. 489.0 to 493.0 nm (Na2CO3 sol.)
Absorbance (E1%1cm) min. 1900 (Na2CO3 sol., 489.0 to 493.0 nm)