3-Fluorosalicylic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

F0845-5G F0845-1G
TCIAF0845-5GEA 2425 SEK
TCIAF0845-5G TCIAF0845-1G
3-Fluorosalicylic acid ≥98.0% (by GC, titration analysis)
3-Fluorosalicylic acid
Formel: C₇H₅FO₃
MW: 156,11 g/mol
MDL Number: MFCD00153167
CAS nummer: 341-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28d614d7fb733ed17338e68fc27b953726b69ee5&vid=28d614d7fb733ed17338e68fc27b953726b69ee5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR