4,4,4-Trifluoro-1-butanol ≥98.0%

Leverantör: TCI EUROPE
Varning

T2781-5G T2781-1G
TCIAT2781-5GEA 1200 SEK
TCIAT2781-5G TCIAT2781-1G
4,4,4-Trifluoro-1-butanol ≥98.0%
4,4,4-Trifluoro-1-butanol
Formel: C₄H₇F₃O
MW: 128,09 g/mol
Kokpunkt: 125°C
Densitet: 1,217 g/cm³ (25 °C)
Flampunkt: 52°C
MDL Number: MFCD00041483
CAS nummer: 461-18-7
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d70846f1c467f6ee70a1044fa7701e94955ab7&vid=f2d70846f1c467f6ee70a1044fa7701e94955ab7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR