Gentamicin sulphate
Leverantör: TCI EUROPE
Fara
Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
G0383-5G G0383-1G
TCIAG0383-5GEA 1725 SEK
TCIAG0383-5G TCIAG0383-1G
Gentamicin sulphate
Gentamicin sulphate
Smältpunkt: 218…237 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00270181
CAS nummer: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee71ebe0c72b3435480b5a86fcda97a6cc0d93e3&vid=ee71ebe0c72b3435480b5a86fcda97a6cc0d93e3&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Total Nitrogen 8.0 to 10.0 %
Specific rotation [a]20/D +107.0 to +121.0 deg (C=1, H2O) (calcd.on anh.substance)
Water max. 18.0 %