Picolinoyl chloride hydrochloride ≥93.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Pyridine-2-carbonyl chloride hydrochloride
P0619-25G
P0619-5G
TCIAP0619-25GEA
3775
SEK
TCIAP0619-25G
TCIAP0619-5G
Picolinoyl chloride hydrochloride ≥93.0%
Picolinoyl chloride hydrochloride
Formel:
C₆H₅Cl₂NO MW: 178,0160 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00060193 CAS nummer: 39901-94-5 UN: 3261 ADR: 8,II |
Specifikation testresultat
Appearance | White to Gray to Red powder to crystal |
Purity (Argentmetric Titration) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...