1,2,5-Trimethylpyrrole ≥98.0%

Leverantör: TCI EUROPE
Varning

T1939-25ML T1939-5ML
TCIAT1939-25MLEA 1400 SEK
TCIAT1939-25ML TCIAT1939-5ML
1,2,5-Trimethylpyrrole ≥98.0%
1,2,5-Trimethylpyrrole
Formel: C₇H₁₁N
MW: 109,1689 g/mol
Flampunkt: 52°C
MDL Number: MFCD00003090
CAS nummer: 930-87-0
EINECS: 213-225-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7c8607964d6385baab845b66a772ec3a38589c5&vid=d7c8607964d6385baab845b66a772ec3a38589c5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR