4-Benzoylpiperidine hydrochloride 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Phenyl(piperidin-4-yl)methanone hydrochloride
232860010 232860100
ACRO232860010EA 725 SEK
ACRO232860010 ACRO232860100
4-Benzoylpiperidine hydrochloride 98%
4-Benzoylpiperidine hydrochloride
Formel: C₁₂H₁₆ClNO
MW: 225.7200 g/mol
Smältpunkt: 226…227 °C
Storage Temperature: Omgivande
MDL Number: MFCD00066982
CAS nummer: 25519-80-6
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %