5,5'-Dimethyl-2,2'-bipyridyl ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline
H26378.06 H26378.14
H26378.06EA 1825 SEK
H26378.06 H26378.14
5,5'-Dimethyl-2,2'-bipyridyl ≥98%
5,5'-Dimethyl-2,2'-bipyridyl
Purity: 98% CAS No.: 1762-34-1 Molecular Formula: C12H12N2 Formula Weight: 184.24 Melting Point: 114-117° Boiling Point: 140°/3mm MDL No.: MFCD01740554
Formel: C₁₂H₁₂N₂
Kokpunkt: 140 °C (3 mmHg)
Smältpunkt: 114…117 °C
Storage Temperature: Omgivande
MDL Number: MFCD01740554
CAS nummer: 1762-34-1